Assay Details
| Gene Symbol: ddx39b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200943.3 | NP_957237.1 | 6 - 7 | 841 | 62 | |
| GenBank mRNA | BC055240.1 | - | 6 - 7 | 812 | 62 | |
| EH457748.1 | - | 6 - 7 | 845 | 62 | ||
More Information
Set Membership: |
|

ddx39b
Genomic Map