Assay Details
| Gene Symbol: pdcd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200950.1 | NP_957244.1 | 1 - 2 | 151 | 61 | |
| GenBank mRNA | BC053162.1 | - | 1 - 2 | 151 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pdcd6| Gene Symbol: pdcd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200950.1 | NP_957244.1 | 1 - 2 | 151 | 61 | |
| GenBank mRNA | BC053162.1 | - | 1 - 2 | 151 | 61 | |
Set Membership: |
|