Assay Details
| Gene Symbol: cpdp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201064.1 | NP_957358.1 | 2 - 3 | 369 | 62 | |
| XM_005168414.4 | XP_005168471.1 | 2 - 3 | 434 | 62 | ||
| XM_005168415.4 | XP_005168472.1 | 2 - 3 | 455 | 62 | ||
| XM_021479428.1 | XP_021335103.1 | 2 - 3 | 440 | 62 | ||
| XM_021479429.1 | XP_021335104.1 | 2 - 3 | 416 | 62 | ||
| XM_021479435.1 | XP_021335110.1 | 2 - 3 | 434 | 62 | ||
| XM_021479436.1 | XP_021335111.1 | 2 - 3 | 435 | 62 | ||
| XM_021479440.1 | XP_021335115.1 | 2 - 3 | 435 | 62 | ||
| GenBank mRNA | BC054710.1 | - | 2 - 3 | 369 | 62 | |
More Information
Set Membership: |
|

cpdp
Genomic Map