Assay Details
| Gene Symbol: nap1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201318.1 | NP_958475.1 | 9 - 10 | 877 | 59 | |
| XM_005171666.3 | XP_005171723.1 | 9 - 10 | 1032 | 59 | ||
| XM_005171667.3 | XP_005171724.1 | 9 - 10 | 1020 | 59 | ||
| XM_021474937.1 | XP_021330612.1 | 9 - 10 | 1032 | 59 | ||
| GenBank mRNA | BC044152.1 | - | 9 - 10 | 877 | 59 | |
| BC165826.1 | - | 7 - 8 | 777 | 59 | ||
More Information
Set Membership: |
|

nap1l1
Genomic Map