Assay Details
| Gene Symbol: hspa9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201326.2 | NP_958483.2 | 16 - 17 | 1992 | 66 | |
| GenBank mRNA | BC044175.1 | - | 16 - 17 | 2094 | 66 | |
| BC083504.1 | - | 16 - 17 | 1992 | 66 | ||
| BC164566.1 | - | 16 - 17 | 2002 | 66 | ||
More Information
Set Membership: |
|

hspa9
Genomic Map