Assay Details
| Gene Symbol: tppp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201335.2 | NP_958492.2 | 3 - 4 | 465 | 60 | |
| GenBank mRNA | BC062530.1 | - | 3 - 4 | 437 | 60 | |
| EH455616.1 | - | 3 - 4 | 497 | 60 | ||
More Information
Set Membership: |
|

tppp3
Genomic Map