Assay Details
| Gene Symbol: nono | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201579.1 | NP_963873.1 | 3 - 4 | 426 | 74 | |
| XM_005165105.4 | XP_005165162.1 | 3 - 4 | 808 | 74 | ||
| XM_021476008.1 | XP_021331683.1 | 2 - 3 | 339 | 74 | ||
| GenBank mRNA | BC046880.1 | - | 3 - 4 | 426 | 74 | |
More Information
Set Membership: |
|

nono
Genomic Map