Assay Details
| Gene Symbol: nqo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204272.2 | NP_001191201.1 | 6 - 7 | 823 | 71 | |
| NM_205542.3 | NP_991105.1 | 6 - 7 | 781 | 71 | ||
| GenBank mRNA | BC065622.1 | - | 6 - 7 | 713 | 71 | |
| EB941347.1 | - | 2 - 3 | 198 | 71 | ||
| EH498485.1 | - | 6 - 7 | 806 | 71 | ||
| EH536561.1 | - | 6 - 7 | 812 | 71 | ||
More Information
Set Membership: |
|

nqo1
Genomic Map