Assay Details
| Gene Symbol: atad3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205703.1 | NP_991266.1 | 1686 | 110 | ||
| GenBank mRNA | BC065962.1 | - | 1686 | 110 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

atad3| Gene Symbol: atad3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205703.1 | NP_991266.1 | 1686 | 110 | ||
| GenBank mRNA | BC065962.1 | - | 1686 | 110 | ||
Set Membership: |
|