Assay Details
| Gene Symbol: fbxo25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205724.1 | NP_991287.1 | 7 - 8 | 710 | 62 | |
| XM_005162395.4 | XP_005162452.1 | 7 - 8 | 789 | 62 | ||
| XM_021469771.1 | XP_021325446.1 | 7 - 8 | 699 | 62 | ||
| GenBank mRNA | AY398345.1 | - | 7 - 8 | 710 | 62 | |
| BC078303.1 | - | 7 - 8 | 780 | 62 | ||
More Information
Set Membership: |
|

fbxo25
Genomic Map