Assay Details
| Gene Symbol: cnot6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212660.2 | NP_997825.1 | 5 - 6 | 593 | 66 | |
| XM_009295585.3 | XP_009293860.1 | 5 - 6 | 578 | 66 | ||
| GenBank mRNA | BC058309.1 | - | 5 - 6 | 516 | 66 | |
| CT690778.2 | - | 5 - 6 | 547 | 66 | ||
More Information
Set Membership: |
|

cnot6a
Genomic Map