Assay Details
| Gene Symbol: mpx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001351837.1 | NP_001338766.1 | 11 - 12 | 1810 | 67 | |
| NM_212779.2 | NP_997944.2 | 11 - 12 | 1810 | 67 | ||
| XM_021479113.1 | XP_021334788.1 | 11 - 12 | 1873 | 67 | ||
| GenBank mRNA | AF349034.1 | - | 11 - 12 | 1811 | 67 | |
| AF378824.1 | - | 8 - 9 | 1487 | 67 | ||
| AF378825.1 | - | 2 - 3 | 277 | 67 | ||
| AF378826.1 | - | 2 - 3 | 296 | 67 | ||
| BC056287.1 | - | 11 - 12 | 1786 | 67 | ||
| BC068379.1 | - | 11 - 12 | 1785 | 67 | ||
More Information
Set Membership: |
|

mpx
Genomic Map