Assay Details
| Gene Symbol: mpx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF349034.1 | - | 14 - 15 | 2291 | 99 | |
| AF378824.1 | - | 11 - 12 | 1967 | 99 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mpx| Gene Symbol: mpx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF349034.1 | - | 14 - 15 | 2291 | 99 | |
| AF378824.1 | - | 11 - 12 | 1967 | 99 | ||
Set Membership: |
|