Assay Details
| Gene Symbol: mpx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001351837.1 | NP_001338766.1 | 7 - 8 | 871 | 56 | |
| NM_212779.2 | NP_997944.2 | 7 - 8 | 871 | 56 | ||
| XM_021479113.1 | XP_021334788.1 | 7 - 8 | 934 | 56 | ||
| GenBank mRNA | AF349034.1 | - | 7 - 8 | 872 | 56 | |
| AF378824.1 | - | 4 - 5 | 548 | 56 | ||
| BC056287.1 | - | 7 - 8 | 847 | 56 | ||
| BC068379.1 | - | 7 - 8 | 846 | 56 | ||
More Information
Set Membership: |
|

mpx
Genomic Map