Assay Details
| Gene Symbol: mpx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF349034.1 | - | 8 - 9 | 1232 | 84 | |
| BC068379.1 | - | 8 - 9 | 1206 | 84 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mpx| Gene Symbol: mpx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF349034.1 | - | 8 - 9 | 1232 | 84 | |
| BC068379.1 | - | 8 - 9 | 1206 | 84 | ||
Set Membership: |
|