Assay Details
| Gene Symbol: myd88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212814.2 | NP_997979.2 | 3 - 4 | 748 | 90 | |
| GenBank mRNA | AY388401.1 | - | 3 - 4 | 612 | 90 | |
| AY389463.1 | - | 1 - 2 | 193 | 90 | ||
| BC083419.1 | - | 3 - 4 | 748 | 90 | ||
| BC164642.1 | - | 3 - 4 | 646 | 90 | ||
| DQ100359.1 | - | 3 - 4 | 668 | 90 | ||
More Information
Set Membership: |
|

myd88
Genomic Map