Assay Details
| Gene Symbol: esrrga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212954.1 | NP_998119.1 | 4 - 5 | 798 | 53 | |
| XM_005158771.4 | XP_005158828.1 | 6 - 7 | 1281 | 53 | ||
| XM_009293126.3 | XP_009291401.1 | 6 - 7 | 1261 | 53 | ||
| XM_021467129.1 | XP_021322804.1 | 6 - 7 | 1294 | 53 | ||
| XM_021467130.1 | XP_021322805.1 | 6 - 7 | 1274 | 53 | ||
| XM_021467131.1 | XP_021322806.1 | 6 - 7 | 1298 | 53 | ||
| XM_021467132.1 | XP_021322807.1 | 7 - 8 | 1132 | 53 | ||
| XM_021467133.1 | XP_021322808.1 | 5 - 6 | 1166 | 53 | ||
| XM_021467134.1 | XP_021322809.1 | 6 - 7 | 1003 | 53 | ||
| XM_021467135.1 | XP_021322810.1 | 5 - 6 | 866 | 53 | ||
| GenBank mRNA | AY556397.1 | - | 4 - 5 | 798 | 53 | |
More Information
Set Membership: |
|

esrrga
Genomic Map