Assay Details
| Gene Symbol: dusp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213067.1 | NP_998232.1 | 2 - 3 | 697 | 84 | |
| GenBank mRNA | BC045494.1 | - | 2 - 3 | 697 | 84 | |
| BC071309.1 | - | 2 - 3 | 702 | 84 | ||
More Information
Set Membership: |
|

dusp1
Genomic Map