Assay Details
| Gene Symbol: acbd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213095.1 | NP_998260.1 | 4 - 5 | 404 | 64 | |
| GenBank mRNA | BC068326.1 | - | 4 - 5 | 404 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

acbd4| Gene Symbol: acbd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213095.1 | NP_998260.1 | 4 - 5 | 404 | 64 | |
| GenBank mRNA | BC068326.1 | - | 4 - 5 | 404 | 64 | |
Set Membership: |
|