Assay Details
| Gene Symbol: pcbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213117.1 | NP_998282.1 | 3 - 4 | 300 | 62 | |
| XM_009296477.3 | XP_009294752.1 | 3 - 4 | 361 | 62 | ||
| XM_009296478.3 | XP_009294753.1 | 3 - 4 | 361 | 62 | ||
| XM_017353108.2 | XP_017208597.1 | 3 - 4 | 361 | 62 | ||
| XM_017353109.2 | XP_017208598.1 | 3 - 4 | 361 | 62 | ||
| GenBank mRNA | BC056323.1 | - | 3 - 4 | 300 | 62 | |
| BC065955.1 | - | 3 - 4 | 316 | 62 | ||
More Information
Set Membership: |
|

pcbp4
Genomic Map