Assay Details
| Gene Symbol: amfra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213163.2 | NP_998328.2 | 11 - 12 | 1502 | 60 | |
| GenBank mRNA | BC057411.1 | - | 10 - 11 | 1490 | 60 | |
More Information
Set Membership: |
|

amfra
Genomic Map