Assay Details
| Gene Symbol: amfra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213163.2 | NP_998328.2 | 13 - 14 | 1836 | 101 | |
| GenBank mRNA | BC057411.1 | - | 12 - 13 | 1824 | 101 | |
More Information
Set Membership: |
|

amfra
Genomic Map