Assay Details
| Gene Symbol: amfra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213163.2 | NP_998328.2 | 6 - 7 | 839 | 91 | |
| GenBank mRNA | BC057411.1 | - | 5 - 6 | 827 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

amfra| Gene Symbol: amfra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213163.2 | NP_998328.2 | 6 - 7 | 839 | 91 | |
| GenBank mRNA | BC057411.1 | - | 5 - 6 | 827 | 91 | |
Set Membership: |
|