Assay Details
| Gene Symbol: kyat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213359.2 | NP_998524.2 | 10 - 11 | 1087 | 86 | |
| XM_005171922.4 | XP_005171979.1 | 10 - 11 | 1187 | 86 | ||
| XM_005171923.4 | XP_005171980.1 | 10 - 11 | 1175 | 86 | ||
| GenBank mRNA | BC055182.1 | - | 10 - 11 | 1007 | 86 | |
| DT059481.1 | - | 3 - 4 | 279 | 86 | ||
More Information
Set Membership: |
|

kyat1
Genomic Map