Assay Details
| Gene Symbol: cdk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213406.1 | NP_998571.1 | 3 - 4 | 478 | 62 | |
| GenBank mRNA | BC049499.1 | - | 3 - 4 | 478 | 62 | |
| BC062836.1 | - | 3 - 4 | 514 | 62 | ||
More Information
Set Membership: |
|

cdk2
Genomic Map