Assay Details
| Gene Symbol: abce1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213051.2 | NP_998216.2 | 12 - 13 | 1386 | 66 | |
| GenBank mRNA | AY391404.1 | - | 12 - 13 | 1366 | 66 | |
| BC045882.1 | - | 12 - 13 | 1392 | 66 | ||
| BC095592.1 | - | 12 - 13 | 1392 | 66 | ||
| BC164863.1 | - | 11 - 12 | 1238 | 66 | ||
| CT630191.2 | - | 4 - 5 | 501 | 66 | ||
More Information
Set Membership: |
|

abce1
Genomic Map