Assay Details
| Gene Symbol: cdkn1ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002040.1 | NP_001002040.1 | 2 - 3 | 693 | 76 | |
| GenBank mRNA | BC071317.1 | - | 2 - 3 | 693 | 76 | |
| BC153451.1 | - | 4 - 5 | 1091 | 76 | ||
More Information
Set Membership: |
|

cdkn1ca
Genomic Map