Assay Details
| Gene Symbol: smpx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002185.1 | NP_001002185.1 | 3 - 4 | 238 | 82 | |
| XM_005162767.4 | XP_005162824.1 | 3 - 4 | 267 | 82 | ||
| XM_009297459.3 | XP_009295734.1 | 3 - 4 | 234 | 82 | ||
| GenBank mRNA | BC074039.1 | - | 3 - 4 | 238 | 82 | |
| BC164228.1 | - | 164 | 82 | |||
More Information
Set Membership: |
|

smpx
Genomic Map