Assay Details
| Gene Symbol: rabepk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002209.1 | NP_001002209.1 | 5 - 6 | 999 | 64 | |
| XM_005171726.4 | XP_005171783.1 | 6 - 7 | 797 | 64 | ||
| GenBank mRNA | BC072705.1 | - | 5 - 6 | 999 | 64 | |
| BC164002.1 | - | 5 - 6 | 737 | 64 | ||
More Information
Set Membership: |
|

rabepk
Genomic Map