Assay Details
| Gene Symbol: spag6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002210.1 | NP_001002210.1 | 5 - 6 | 790 | 68 | |
| GenBank mRNA | BC074072.1 | - | 5 - 6 | 790 | 68 | |
| BC165187.1 | - | 5 - 6 | 710 | 68 | ||
More Information
Set Membership: |
|

spag6
Genomic Map