Assay Details
| Gene Symbol: eef1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002371.1 | NP_001002371.1 | 5 - 6 | 921 | 106 | |
| GenBank mRNA | BC075885.1 | - | 5 - 6 | 921 | 106 | |
More Information
Set Membership: |
|

eef1a2
Genomic Map