Assay Details
| Gene Symbol: rab40b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002524.1 | NP_001002524.2 | 3 - 4 | 710 | 73 | |
| GenBank mRNA | BC076375.1 | - | 3 - 4 | 710 | 73 | |
More Information
Set Membership: |
|

rab40b
Genomic Map