Assay Details
Gene Symbol: rab40b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002524.1 | NP_001002524.2 | 4 - 5 | 788 | 116 | |
GenBank mRNA | BC076375.1 | - | 4 - 5 | 788 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
