Assay Details
| Gene Symbol: sat2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002554.2 | NP_001002554.2 | 1 - 2 | 208 | 117 | |
| NM_001362464.1 | NP_001349393.1 | 1 - 2 | 208 | 117 | ||
| GenBank mRNA | BC076248.1 | - | 1 - 2 | 169 | 117 | |
| EH462485.1 | - | 3 - 4 | 519 | 117 | ||
| EH498050.1 | - | 1 - 2 | 215 | 117 | ||
More Information
Set Membership: |
|

sat2b
Genomic Map