Assay Details
| Gene Symbol: sat2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002554.2 | NP_001002554.2 | 5 - 6 | 490 | 82 | |
| GenBank mRNA | BC076248.1 | - | 5 - 6 | 451 | 82 | |
| EH498050.1 | - | 5 - 6 | 497 | 82 | ||
More Information
Set Membership: |
|

sat2b
Genomic Map