Assay Details
| Gene Symbol: crlf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002650.1 | NP_001002650.1 | 7 - 8 | 1390 | 103 | |
| XM_017358225.2 | XP_017213714.1 | 8 - 9 | 1722 | 103 | ||
| GenBank mRNA | BC076526.1 | - | 7 - 8 | 1420 | 103 | |
More Information
Set Membership: |
|

crlf1a
Genomic Map