Assay Details
| Gene Symbol: denr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002697.2 | NP_001002697.1 | 926 | 104 | ||
| GenBank mRNA | BC076117.1 | - | 587 | 104 | ||
| BC095896.1 | - | 926 | 104 | |||
More Information
Set Membership: |
|

denr
Genomic Map