Assay Details
| Gene Symbol: adprh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003453.2 | NP_001003453.1 | 7 - 8 | 1209 | 72 | |
| GenBank mRNA | BC078399.1 | - | 7 - 8 | 1055 | 72 | |
| BC164170.1 | - | 6 - 7 | 947 | 72 | ||
More Information
Set Membership: |
|

adprh
Genomic Map