Assay Details
| Gene Symbol: prkacab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003470.1 | NP_001003470.1 | 5 - 6 | 836 | 94 | |
| GenBank mRNA | BC078343.1 | - | 5 - 6 | 836 | 94 | |
More Information
Set Membership: |
|

prkacab
Genomic Map