Assay Details
| Gene Symbol: cux1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003514.1 | NP_001003514.1 | 8 - 9 | 712 | 85 | |
| GenBank mRNA | BC075748.1 | - | 8 - 9 | 712 | 85 | |
More Information
Set Membership: |
|

cux1b
Genomic Map