Assay Details
| Gene Symbol: rpp21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003530.2 | NP_001003530.1 | 1 - 2 | 153 | 79 | |
| GenBank mRNA | BC078300.1 | - | 1 - 2 | 157 | 79 | |
More Information
Set Membership: |
|

rpp21
Genomic Map