Assay Details
| Gene Symbol: nek7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003617.1 | NP_001003617.1 | 6 - 7 | 738 | 56 | |
| GenBank mRNA | BC077138.1 | - | 6 - 7 | 703 | 56 | |
| BC081618.1 | - | 6 - 7 | 738 | 56 | ||
More Information
Set Membership: |
|

nek7
Genomic Map