Assay Details
| Gene Symbol: eif3ha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003763.2 | NP_001003763.1 | 2 - 3 | 313 | 66 | |
| GenBank mRNA | BC079514.1 | - | 2 - 3 | 306 | 66 | |
| EH509704.1 | - | 2 - 3 | 307 | 66 | ||
More Information
Set Membership: |
|

eif3ha
Genomic Map