Assay Details
| Gene Symbol: eif3ha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003763.2 | NP_001003763.1 | 5 - 6 | 724 | 68 | |
| GenBank mRNA | BC079514.1 | - | 5 - 6 | 717 | 68 | |
| BI473261.1 | - | 3 - 4 | 521 | 68 | ||
| EH509704.1 | - | 5 - 6 | 718 | 68 | ||
More Information
Set Membership: |
|

eif3ha
Genomic Map