Assay Details
| Gene Symbol: eif3ha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003763.2 | NP_001003763.1 | 7 - 8 | 984 | 144 | |
| GenBank mRNA | BC079514.1 | - | 7 - 8 | 977 | 144 | |
| BI473261.1 | - | 1 - 2 | 261 | 144 | ||
More Information
Set Membership: |
|

eif3ha
Genomic Map