Assay Details
| Gene Symbol: atp5po | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003843.1 | NP_001003843.1 | 5 - 6 | 443 | 69 | |
| GenBank mRNA | AY648828.1 | - | 5 - 6 | 443 | 69 | |
| BC095099.1 | - | 5 - 6 | 460 | 69 | ||
| BC164303.1 | - | 5 - 6 | 464 | 69 | ||
More Information
Set Membership: |
|

atp5po
Genomic Map