Assay Details
| Gene Symbol: psmb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003889.1 | NP_001003889.1 | 3 - 4 | 326 | 73 | |
| GenBank mRNA | AY648806.1 | - | 3 - 4 | 326 | 73 | |
| BC085580.1 | - | 3 - 4 | 321 | 73 | ||
| BC165077.1 | - | 3 - 4 | 316 | 73 | ||
More Information
Set Membership: |
|

psmb1
Genomic Map