Assay Details
| Gene Symbol: stat5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003984.1 | NP_001003984.1 | 16 - 17 | 2212 | 71 | |
| GenBank mRNA | AY501427.1 | - | 16 - 17 | 2212 | 71 | |
More Information
Set Membership: |
|

stat5b
Genomic Map