Assay Details
| Gene Symbol: ofd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004496.2 | NP_001004496.2 | 17 - 18 | 2382 | 62 | |
| NM_001319208.1 | NP_001306137.1 | 17 - 18 | 2385 | 62 | ||
| XM_009305014.3 | XP_009303289.1 | 17 - 18 | 2535 | 62 | ||
| GenBank mRNA | AJ534306.1 | - | 17 - 18 | 2379 | 62 | |
| BC083518.1 | - | 17 - 18 | 2385 | 62 | ||
| BC096786.1 | - | 17 - 18 | 2367 | 62 | ||
| BC164049.1 | - | 17 - 18 | 2236 | 62 | ||
More Information
Set Membership: |
|

ofd1
Genomic Map