Assay Details
| Gene Symbol: bmpr1ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004585.1 | NP_001004585.1 | 10 - 11 | 1622 | 71 | |
| GenBank mRNA | BC081626.1 | - | 10 - 11 | 1622 | 71 | |
More Information
Set Membership: |
|

bmpr1ab
Genomic Map